Learn More
Dräger™ CMS Chips: Sulfur Dioxide
Suitable for 10 measurements of one gas
Supplier:  Dräger™ 6406110
 View more versions of this product
Catalog No. 18-999-1931
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
5 to 20mg/L
40 to 300 sec.
5°C to 30°C
Sulfur Dioxide CMS Chip
0.4 to 10
41°F to 86°F
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.