Learn More
Dräger™ CMS Chips: Vinyl Chloride
Catalog No. 19034577 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
0.3 to 10
10 to 250
Response Time:
30 to 420 sec.
45 to 100 sec.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Response Time
19-034-577 0.3 to 10 30 to 420 sec.
19-034-578 10 to 250 45 to 100 sec.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-034-577 Supplier Dräger™ Supplier No. 6406170
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Vinyl Chloride CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 0.3 to 10
Response Time 30 to 420 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C