Learn More
Dräger™ Individual Haz Mat Simultaneous Test Sets
For simultaneous testing of five different gases
$159.18
Specifications
For Use With (Application) For simultaneous testing of five different gases
View More Specs
Alert:
No other type of tube should be substituted with these kits.
Requires: Require adapter and a Dräger pump (sold separately) for the semi-quantitative determination of inorganic gases and organic vapors.
Products 3
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
18-556R
View Documents
Dräger™
8101735
Simultaneous Test Set I
Pack of 5 for $159.18
Only null left
Add to Cart
18-556S
View Documents
Dräger™
8101736
Simultaneous Test Set II
Pack of 5 for $159.18
Only null left
Add to Cart
18-556T
missing translation for 'viewDocuments'
Dräger™
8101770
Simultaneous Test Set III
Pack of 5 for $159.18
Only null left
Add to Cart
Description
Description
Test Set I. Includes detector tubes for hydrogen chloride, hydrogen cyanide, carbon monoxide, ammonia, and nitrous fumes.
Test Set II. Includes detector tubes for sulfur dioxide, chlorine, hydrogen sulfide, carbon dioxide, and phosgene.
Test Set III. Includes detector tubes for alcohols, aliphatic hydrocarbons, aromatics, chlorinated hydrocarbons, and ketones.
Order Info
Packs of 10 test sets.
Specifications
Specifications
For simultaneous testing of five different gases
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.