Learn More
Description
Includes
Alligator clip for secure attachment
These hardy sensors can be used in extreme environmental conditions without triggering false alarms.
- Compact, robust and ergonomic design with intuitive two-button operation
- Ideal for personal monitoring of industrial oxygen, carbon monoxide, or hydrogen sulfide
- Large, language-free backlit display continuously indicates gas concentration while simultaneously displaying the corresponding measurement unit
- Equipped with a sensor that allows gas intake from both the top and front of the unit to ensure continuous measurement, even if a gas inlet is covered
- Triple alarms for exceptional safety—clear, multi-toned signal, a bright, flashing 360° LED on top and bottom, and vibration
- Store up to 60 events, along with dates and times; infrared interface enables data to be loaded onto a PC for analysis (requires connecting cradle or E-Cal System and software, sold separately)
- When bump mode is activated, the unit automatically reminds users when a bump test or calibration is required
- Operates in a temperature range from -22° to +122°F (-30° to +50°C); relative humidity, 10 to 90%
Order Info
Dimensions: 0.8L x 2.5W x 3.3 in. H (2.0 x 6.4 x 8.4cm). Weight: 3.8 oz. (106g).
Certifications
IP65 rated for ingresss protection. UL Classified as Intrinsically Safe for use in Class I, Division 1, Group A, B, C, D, E, F, G hazardous locations, Temp Code T4; ATEX, I/II 1/2 G EEx ia I/IIC, T4. CE mark.
Compliance
Complies with EMC Directive 89/336/EEC.
Specifications
Specifications
| Alarm Types | Audible, visual, vibrating |
| Battery Life | 8 hr. |
| Depth (English) | 1 in. |
| Depth (Metric) | 2.5 cm |
| Components Measured | Carbon Monoxide |
| Height (English) | 2.5 in. |
| Height (Metric) | 6.4 cm |
| Humidity Range | 10 to 90% |
| Includes | Alligator clip |
| IP Rating | IP65 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.