Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Pac™ 5500 Single-Gas Detector

Catalog No. 17985684 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Components Measured:
Carbon Monoxide
Oxygen
Measurement Range:
0 to 25%
0 to 500
Response Time:
10 sec.
15 sec.
Includes:
Alligator clip
Blue keypad overlay, Alligator clip
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Measurement Range Response Time Includes
17-985-684 Oxygen 0 to 25% 10 sec. Blue keypad overlay, Alligator clip
17-985-680 Carbon Monoxide 0 to 500 15 sec. Alligator clip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-985-684 Supplier Dräger™ Supplier No. 8322013

Includes

Alligator clip for secure attachment

Unlimited instrument life

These hardy sensors can be used in extreme environmental conditions without triggering false alarms.

  • Compact, robust and ergonomic design with intuitive two-button operation
  • Ideal for personal monitoring of industrial oxygen, carbon monoxide, or hydrogen sulfide
  • Large, language-free backlit display continuously indicates gas concentration while simultaneously displaying the corresponding measurement unit
  • Equipped with a sensor that allows gas intake from both the top and front of the unit to ensure continuous measurement, even if a gas inlet is covered
  • Triple alarms for exceptional safety—clear, multi-toned signal, a bright, flashing 360° LED on top and bottom, and vibration
  • Store up to 60 events, along with dates and times; infrared interface enables data to be loaded onto a PC for analysis (requires connecting cradle or E-Cal System and software, sold separately)
  • When bump mode is activated, the unit automatically reminds users when a bump test or calibration is required
  • Operates in a temperature range from -22° to +122°F (-30° to +50°C); relative humidity, 10 to 90%

Order Info

Dimensions: 0.8L x 2.5W x 3.3 in. H (2.0 x 6.4 x 8.4cm). Weight: 3.8 oz. (106g).

Certifications

IP65 rated for ingresss protection. UL Classified as Intrinsically Safe for use in Class I, Division 1, Group A, B, C, D, E, F, G hazardous locations, Temp Code T4; ATEX, I/II 1/2 G EEx ia I/IIC, T4. CE mark.

Compliance

Complies with EMC Directive 89/336/EEC.

Specifications

Alarm Types Audible, visual, vibrating
Battery Life 8 hr.
Depth (English) 1 in.
Depth (Metric) 2.5 cm
Components Measured Oxygen
Height (English) 2.5 in.
Height (Metric) 6.4 cm
Humidity Range 10 to 90%
Includes Blue keypad overlay, Alligator clip
IP Rating IP65
Measurement Range 0 to 25%
Pressure Range 700 to 1300hpa
Resolution 0.1%
Response Time 10 sec.
Temperature Range (English) -20°F to +120°F
Temperature Range (Metric) -30°C to +50°C
Weight (English) 3.8 oz.
Weight (Metric) 106g
Width (English) 3.3 in.
Width (Metric) 8.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.