Learn More
Dräger™ X-am™ 2500 Multi-Gas Detection
Catalog No. 17985648 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Components Measured:
Ex, Oxygen, Carbon Monoxide, Hydrogen Sulfide
Catalog No. Components Measured
17-985-648 Ex, Oxygen, Carbon Monoxide, Hydrogen Sulfide
1 options
Catalog No. 17-985-648 Supplier Dräger™ Supplier No. 4542261
Instrument provides personal protection in industrial, mining, refinery, and confined space applications. Lightweight and easy to operate, it shows all readings at once on the large LCD and stores peak values, which are easily reset.
  • Suitable for use in explosion zone 0 areas
  • IP67 water- and dust-resistant
  • X-dock compatible
Specifications
Battery Type Nickel-Metal Hydride, 12 hr.
Components Measured Ex, Oxygen, Carbon Monoxide, Hydrogen Sulfide
Type Gas Detector Kit
Includes Stated Sensors, Training CD, Nickel Metal Hydride Battery Pack, Charging Cradle and 110V Charging Plug
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.