Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Short-Term Detector Tubes: Sulfur Dioxide

Catalog No. 17985301 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
0.1 to 3
0.5 to 25
1 to 25
20 to 2000
50 to 8000
Tube Type:
Sulfur dioxide 0.1/a
Sulfur dioxide 0.5/a2
Sulfur dioxide 1/a
Sulfur dioxide 20/a
Sulfur dioxide 50/b
Response Time:
20 min.
3 min.
3 to 6 min.
Not Applicable
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Tube Type Response Time
17-985-301 20 to 2000 Sulfur dioxide 20/a 3 min.
17-986-71C 50 to 8000 Sulfur dioxide 50/b Not Applicable
17-985-299 0.5 to 25 Sulfur dioxide 0.5/a2 3 to 6 min.
17-985-300 1 to 25 Sulfur dioxide 1/a 3 min.
17-985-298 0.1 to 3 Sulfur dioxide 0.1/a 20 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-985-301 Supplier Dräger™ Supplier No. CH24201
Add to Cart
Add to Cart

Detector tubes

Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.

Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.

Specifications

Components Measured Sulfur Dioxide
Humidity Range Less than30mg/L
Measurement Range 20 to 2000
No. of Strokes 10
Quantity 10
Response Time 3 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Tube Type Sulfur dioxide 20/a
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.