Learn More
Dräger™ Short-Term Detector Tubes - De Tube Monostyrene
Catalog No. 17985263 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Components Measured:
Monostyrene 10/a
Monostyrene 10/b
Measurement Range:
10 to 200ppm
10 to 250ppm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Measurement Range
17-985-263 Monostyrene 10/b 10 to 250ppm
17-985-262 Monostyrene 10/a 10 to 200ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-985-263 Supplier Dräger™ Supplier No. 6733141
This item is not returnable. View return policy
  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.
Specifications
Components Measured Monostyrene 10/b
Humidity Range 3 to 15mg/L
Measurement Range 10 to 250ppm
No. of Strokes 20
Quantity 10
Response Time 3 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C