Learn More
Dräger™ Short-Term Detector Tubes - Nitrogen Fumes
For making spot measurments for various gases and vapors
Supplier:  Dräger™ CH31001/DEL
 View more versions of this product
Catalog No. 17-985-273
This item is not returnable. View return policy
Description
Description
  • Glass vial
  • Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
Specifications
Specifications
Nitrous Fumes 2/a
2 to 100ppm
10
50°F to 86°F
30mg/L
5/10
1 to 2 min.
10°C to 30°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.