Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB097460 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB097460 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB097460 Supplier Novus Biologicals Supplier No. NBP297557AF594
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DR1 Polyclonal specifically detects DR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen DR1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias Down-regulator of transcription 1, down-regulator of transcription 1, TBP-binding (negative cofactor 2), NC2, NC2-BETA, Negative cofactor 2-beta, protein Dr1, TATA-binding protein-associated phosphoprotein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DR1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1810
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.