Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DR1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP103329R
Description
DR1 Polyclonal antibody specifically detects DR1 in Human samples. It is validated for Western BlotSpecifications
DR1 | |
Polyclonal | |
Western Blot | |
Down-regulator of transcription 1, down-regulator of transcription 1, TBP-binding (negative cofactor 2), NC2, NC2-BETA, Negative cofactor 2-beta, protein Dr1, TATA-binding protein-associated phosphoprotein | |
The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human down-regulator of transcription 1 using the numbering given in entry NP_001929.1 (GeneID 1810). | |
0.1 mL | |
Stem Cell Markers | |
1810 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction