Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117920
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117920 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117920 Supplier Invitrogen™ Supplier No. PA5117920
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DR1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

This gene encodes a TBP- (TATA box-binding protein) associated phosphoprotein that represses both basal and activated levels of transcription. The encoded protein is phosphorylated in vivo and this phosphorylation affects its interaction with TBP. This protein contains a histone fold motif at the amino terminus, a TBP-binding domain, and a glutamine- and alanine-rich region. The binding of DR1 repressor complexes to TBP-promoter complexes may establish a mechanism in which an altered DNA conformation, together with the formation of higher order complexes, inhibits the assembly of the preinitiation complex and controls the rate of RNA polymerase II transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DR1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene Dr1
Gene Accession No. Q01658
Gene Alias 1700121L09Rik; Down-regulator of transcription 1; down-regulator of transcription 1, TBP-binding (negative cofactor 2); Dr1; Dr1l; NC2; NC2B; NC2beta; NC2-beta; negative cofactor 2; negative cofactor 2 beta; negative cofactor 2-beta; Protein Dr1; similar to down-regulator of transcription 1; TATA-binding protein-associated phosphoprotein; wu:fj15f04; zgc:152914
Gene Symbols Dr1
Host Species Rabbit
Immunogen E. coli-derived Human DR1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1810
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.