Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563106 Supplier Invitrogen™ Supplier No. PA563106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVKEVLQECK TVALKRRKAS SRLENLGIPE EELLRQQQEL FAKARQQQAE LAQQEWLQMQ Q Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a TBP- (TATA box-binding protein) associated phosphoprotein that represses both basal and activated levels of transcription. The encoded protein is phosphorylated in vivo and this phosphorylation affects its interaction with TBP. This protein contains a histone fold motif at the amino terminus, a TBP-binding domain, and a glutamine- and alanine-rich region. The binding of DR1 repressor complexes to TBP-promoter complexes may establish a mechanism in which an altered DNA conformation, together with the formation of higher order complexes, inhibits the assembly of the preinitiation complex and controls the rate of RNA polymerase II transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DR1
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dr1
Gene Accession No. Q01658
Gene Alias 1700121L09Rik; Down-regulator of transcription 1; down-regulator of transcription 1, TBP-binding (negative cofactor 2); Dr1; Dr1l; NC2; NC2B; NC2beta; NC2-beta; negative cofactor 2; negative cofactor 2 beta; negative cofactor 2-beta; Protein Dr1; similar to down-regulator of transcription 1; TATA-binding protein-associated phosphoprotein; wu:fj15f04; zgc:152914
Gene Symbols Dr1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DR1. Recombinant protein control fragment (Product #RP-101741).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1810
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.