Learn More
Specifications
Specifications
| Gene ID (Entrez) | 8718 |
| Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56380 |
| Gene Alias | Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1 |
| Gene Symbol | TNFRSF25 |
| Quantity | 0.1 mg |
| Research Category | Apoptosis, Cell Biology |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
