Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR3/TNFRSF25 Antibody, Novus Biologicals™
SDP

Catalog No. NBP176474 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176474 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176474 Supplier Novus Biologicals Supplier No. NBP176474
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DR3/TNFRSF25 Polyclonal specifically detects DR3/TNFRSF25 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence.

Specifications

Antigen DR3/TNFRSF25
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. AAQ88676
Gene Alias Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1
Gene Symbols TNFRSF25
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the amino terminus of human DR3. The immunogen is located within the first 50 amino acids of DR3. Amino Acid Squence: QGGTRSPRCDCAAGDFH
Molecular Weight of Antigen 59 kDa
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 8718
Test Specificity DR3 has no cross reaction to other death receptors.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.