Learn More
Invitrogen™ DR4 Polyclonal Antibody

Description
Positive test controls include: K-562, SW480, MCF7, HeLa, DU145, Mouse lung, Rat lung. The target is usually found in the following locations: Membrane, Single-pass type I membrane protein. Immunogen sequence: GGDPKCMDRV CFWRLGLLRG PGAEDNAHNE ILSNADSLST FVSEQQMESQ EPADLTGVTV QSPGEAQCLL GPAEAEGSQR RRLLVPANGA DPTETLMLFF DKFANIVPFD SWDQLMRQLD LTKNEIDVVR AGTAGPGDAL YAMLMKWVNK TGRNASIHTL LDALERMEER HAREKIQDLL VDSGKFIYLE DGTGSAVSLE.
Specifications
Specifications
| Antigen | DR4 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.39 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TNFRSF10A |
| Gene Accession No. | O00220 |
| Gene Alias | APO2; CD261; cytotoxic TRAIL receptor; death receptor 4; DR4; MGC9365; sCD261; soluble CD261; soluble TRAIL; sTRAIL; TNF receptor superfamily member 10a; TNF-related apoptosis-inducing ligand receptor 1; TNFRSF10A; TRAIL R1; TRAIL receptor 1; TRAILR1; TRAIL-R1; TRAILR-1; tumor necrosis factor receptor superfamily member 10A; tumor necrosis factor receptor superfamily member 10a variant 2; tumor necrosis factor receptor superfamily, member 10a |
| Gene Symbols | TNFRSF10A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.