Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR6 (CD358) Rat anti-Mouse, Clone: 6B6, MilliporeSigma™

Catalog No. MABC1594100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1594100 100 μL
MABC159425 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1594100 Supplier MilliporeSigma Supplier No. MABC1594100UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

DR6 (CD358) Monoclonal antibody specifically detects DR6 (CD358) in Mouse samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DR6 (CD358)
Applications Western Blot
Classification Monoclonal
Clone 6B6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rat
Immunogen Recombinant fragment corresponding to full-length ectodomain region of mouse DR6 with an N‐terminal CD5 signal peptide and a C‐terminal 2XStrepII tag.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.