Learn More
Description
Specifications
Specifications
| Antigen | DR6 (CD358) |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 6B6 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified rat monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. |
| Host Species | Rat |
| Immunogen | Recombinant fragment corresponding to full-length ectodomain region of mouse DR6 with an N‐terminal CD5 signal peptide and a C‐terminal 2XStrepII tag. |
| Purification Method | Protein G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.