Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DR6/TNFRSF21 Antibody (001), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289313AF647
Description
DR6/TNFRSF21 Monoclonal antibody specifically detects DR6/TNFRSF21 in Human samples. It is validated for ELISASpecifications
DR6/TNFRSF21 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
001 | |
ELISA | |
BM-018, CD358 antigen, Death receptor 6, DR6TNFR-related death receptor 6, MGC31965, tumor necrosis factor receptor superfamily member 21, tumor necrosis factor receptor superfamily, member 21 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human DR6/TNFRSF21 (Met 1-Leu 350; NP_055267.1) . | |
0.1 mL | |
Apoptosis, Cell Biology | |
27242 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction