Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DR6/TNFRSF21 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298249AF750
Description
DR6/TNFRSF21 Polyclonal specifically detects DR6/TNFRSF21 in Mouse samples. It is validated for ELISA.Specifications
DR6/TNFRSF21 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
Alexa Fluor 750 | |
BM-018, CD358 antigen, Death receptor 6, DR6TNFR-related death receptor 6, MGC31965, tumor necrosis factor receptor superfamily member 21, tumor necrosis factor receptor superfamily, member 21 | |
Produced in rabbits immunized with purified, recombinant DR6/TNFRSF21 (NP_055267.1; Met 1-His 349). | |
0.1 mL | |
Apoptosis, Cell Biology | |
27242 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction