Learn More
Dräger™ Short-Term Detector Tubes: Aniline
Catalog No. 17985181 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
0.5 to 10
1 to 20
Tube Type:
Aniline 0.5/a
Aniline 5/a
Humidity Range:
7 to 12mg/L
Less than50mg/L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Tube Type Humidity Range
17-985-181 1 to 20 Aniline 5/a Less than50mg/L
17-985-180 0.5 to 10 Aniline 0.5/a 7 to 12mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-985-181 Supplier Dräger™ Supplier No. CH20401
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Description Detector Tubes
Components Measured Aniline
Humidity Range Less than50mg/L
Measurement Range 1 to 20
No. of Strokes 5 to 25
Quantity 10
Response Time 3 min.
Temperature Range (English) 59°F to 104°F
Temperature Range (Metric) 15° to 40°C
Tube Type Aniline 5/a