Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRAK1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP142692JF549
Description
DRAK1 Polyclonal antibody specifically detects DRAK1 in Human samples. It is validated for Western BlotSpecifications
DRAK1 | |
Polyclonal | |
Western Blot | |
DAP kinase-related apoptosis-inducing protein kinase 1, DRAK1death-associated protein kinase-related 1, EC 2.7.11.1, serine/threonine kinase 17a, serine/threonine kinase 17a (apoptosis-inducing), serine/threonine-protein kinase 17A | |
The immunogen for this product maps to a region between residue 314 and 364 of human DAP kinase-related apoptosis-inducing protein kinase 1 using the numbering given in entry NP_004751.2 (GeneID 9263). | |
0.1 mL | |
Apoptosis, Cell Biology, Signal Transduction | |
9263 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction