Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DRAK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111092
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111092 100 μL
PIPA5110833 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111092 Supplier Invitrogen™ Supplier No. PA5111092
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RTLLVKKPED RATAEECLKH PWLTQSSIQE PSFRMEKALE EANALQEGHS VPEINSDTDK SETKESIVTE Highest antigen sequence indentity to the following orthologs: Mouse - 33%, Rat - 35%.

STK17A (DRAK1) is a serine/threonine kinase localized in the nucleus and involved in apoptosis. Diseases associated with STK17A include Colon Squamous Cell Carcinoma. Among its related pathways are Sweet Taste Signaling. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is STK17B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRAK1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STK17A
Gene Accession No. Q9UEE5
Gene Alias DAP kinase-related apoptosis-inducing protein kinase 1; death-associated protein kinase-related 1; DRAK1; serine/threonine kinase 17a; serine/threonine kinase 17a (apoptosis-inducing); serine/threonine-protein kinase 17A; STK17A
Gene Symbols STK17A
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human DRAK1. Recombinant protein control fragment (Product #RP-97245).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9263
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.