Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRAM Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176759C
This item is not returnable.
View return policy
Description
DRAM Polyclonal antibody specifically detects DRAM in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
DRAM | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
Damage-regulated autophagy modulator, DNA damage-regulated autophagy modulator protein 1, DNA-damage regulated autophagy modulator 1, DRAMFLJ11259 | |
Antibody was raised against a peptide corresponding to 16 amino acids near the amino terminus of human DRAM. The immunogen is located within amino acids 30-80 of DRAM. Amino Acid Sequence: LPYISDTGTTPPESGI | |
0.1 mL | |
Cancer | |
55332 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction