Learn More
Invitrogen™ DRAM Polyclonal Antibody

Description
A suggested positive control is 293 cell lysate. PA5-20335 can be used with blocking peptide PEP-0455.
Specifications
Specifications
| Antigen | DRAM |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | DRAM1 |
| Gene Accession No. | Q8N682, Q9DC58 |
| Gene Alias | 1200002N14Rik; Damage-regulated autophagy modulator; DNA damage regulated autophagy modulator 1; DNA damage-regulated autophagy modulator protein 1; DNA-damage regulated autophagy modulator 1; DRAM; Dram1; FLJ11259; hypothetical protein FLJ11259; zgc:101811 |
| Gene Symbols | DRAM1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.