Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DRAM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113356 Supplier Invitrogen™ Supplier No. PA5113356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DRAM2 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen DRAM2
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DRAM2
Gene Accession No. Q6UX65
Gene Alias 2010305N14Rik; 2610318G18Rik; AI647667; CORD21; damage regulated autophagy modulator 2; DNA damage regulated autophagy modulator 2; DNA damage-regulated autophagy modulator protein 2; DNA-damage regulated autophagy modulator 2; DRAM2; PRO180; PSEC0031; RP5-1180E21.1; TMEM77; Transmembrane protein 77; UNQ154/PRO180; VDNA-damage regulated autophagy modulator 2; WWFQ154
Gene Symbols DRAM2
Host Species Rabbit
Immunogen Recombinant Human DNA damage-regulated autophagy modulator protein 2 protein (182-266AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128338
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.