Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DRAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552348
Description
Immunogen sequence: RIKKIMQTDE EIGKVAAAVP VIISRALELF LESLLKKACQ VTQSRNAKTM TTSHLKQCIE LEQQFDFLKD LVASVPDMQG DGEDNHMDGD KGARRGRKPG SGGRKNGGMG TKSKDKKLSG TDSEQEDESE DTDTDGEEET SQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.

Specifications
DRAP1 | |
Polyclonal | |
Unconjugated | |
DRAP1 | |
2310074H19Rik; DR1 associated protein 1; Dr1 associated protein 1 (negative cofactor 2 alpha); dr1-associated corepressor; Dr1-associated protein 1; DR1-associated protein 1 (negative cofactor 2 alpha); DRAP1; drap1 {ECO:0000250; NC2-alpha; negative cofactor 2 alpha; negative co-factor 2-alpha; negative cofactor 2-alpha; UniProtKB:Q14919} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10589 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14919 | |
DRAP1 | |
Recombinant protein corresponding to Human DRAP1. Recombinant protein control fragment (Product #RP-89425). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction