Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DRAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120562
This item is not returnable.
View return policy
Description
Positive test controls include: U-87MG, Jurkat, 293T, Rat testis. Immunogen sequence: LKKACQVTQS RNAKTMTTSH LKQCIELEQQ FDFLKDLVAS VPDMQGDGED NHMDGDKGAR RGRKPGSGGR KNGGMGTKSK DKKLSGTDSE QEDESEDTDT D.
Specifications
| DRAP1 | |
| Polyclonal | |
| Unconjugated | |
| DRAP1 | |
| 2310074H19Rik; DR1 associated protein 1; Dr1 associated protein 1 (negative cofactor 2 alpha); dr1-associated corepressor; Dr1-associated protein 1; DR1-associated protein 1 (negative cofactor 2 alpha); DRAP1; drap1 {ECO:0000250; NC2-alpha; negative cofactor 2 alpha; negative co-factor 2-alpha; negative cofactor 2-alpha; UniProtKB:Q14919} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10589, 293674, 66556 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.63 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| A0JPP1, Q14919, Q9D6N5 | |
| DRAP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 50-150 of human DRAP1 (NP_006433.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction