Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ DRAQ7™
SDP

Catalog No. NB016659 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μL
1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016659 1000 μL
NB016660 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB016659 Supplier Novus Biologicals™ Supplier No. NBP2811261000UL
Add to Cart
Add to Cart

Specifications

Content And Storage Store at 4°C in the dark. Do not freeze.
Quantity 1000 μL
For Use With (Application) Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.