Learn More
Invitrogen™ DRAQ7 DROP & GO™ Dye
Catalog No. D15107
Encompass_Preferred
Change view
Click to view available options
Quantity:
90 assays
Catalog No. Quantity
D15107 90 assays
1 options
Catalog No. D15107 Supplier Invitrogen™ Supplier No. D15107
  • $497.00
    $372.65 / Each of 1
    Save $124.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart
Used to exclude non-viable cells by flow cytometry
  • DRAQ7 DROP & GO Dye is a bright, easy-to-use fluorescent DNA dye used to exclude non-viable cells by flow cytometry. This dye has excitation maxima at 599/644 nm and emission maxima at 678/697 nm.
  • Stable at room temperature
  • Convenient, ready-to-use dropper-bottle format—no need to dilute, weigh, or pipette
  • No UV excitation and no emission spectral overlap with PE
  • DRAQ7 Dye is a membrane-impermeable dye that rapidly stains the double-stranded DNA (dsDNA) of dead or permeabilized cells. The dye can be excited by wavelengths from 488 to 647 nm, though it is optimally excited by red laser lines. Its emission maxima is at 697 nm when intercalated with dsDNA.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 3 x 2.5 mL
Product Type Dye
Quantity 90 assays
For Research Use Only. Not for use in diagnostic procedures.