Learn More
Invitrogen™ DRAQ7™ Dye
Catalog No. D15105
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 Assays
50 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15105 50 Assays
D15106 200 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15105 Supplier Invitrogen™ Supplier No. D15105
Add to Cart
Edge
Add to Cart
DRAQ7 Dye is a bright, easy-to-use fluorescent DNA dye used to exclude non-viable cells by flow cytometry.
This dye has excitation maxima at 599/644 nm and emission maxima at 678/697 nm.
• Stable at room temperature
• Rapid staining of dead cells without wash steps
• No UV excitation and no emission spectral overlap with PE
DRAQ7 Dye is a membrane-impermeable dye that rapidly stains the double-stranded DNA (dsDNA) of dead or permeabilized cells. The dye can be excited by wavelengths from 488 to 647 nm, though it is optimally excited by red laser lines. Its emission maxima is at 697 nm when intercalated with dsDNA.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
Product Type Dye
Quantity 50 Assays
For Research Use Only. Not for use in diagnostic procedures.