Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Draxin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547816
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547816 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547816 Supplier Invitrogen™ Supplier No. PA547816
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 50% cross-reactivity with recombinant human Draxin is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Chemorepulsive axon guidance protein required for the development of spinal cord and forebrain commissures. Acts as a chemorepulsive guidance protein for commissural axons during development. Able to inhibit or repel neurite outgrowth from dorsal spinal cord. Inhibits the stabilization of cytosolic beta-catenin (CTNNB1) via its interaction with LRP6, thereby acting as an antagonist of Wnt signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Draxin
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene DRAXIN
Gene Accession No. Q6PAL1
Gene Alias 2610109H07Rik; AGPA3119; C1orf187; Dorsal inhibitory axon guidance protein; dorsal repulsive axon guidance protein; Draxin; FLJ34999; MGC117222; Neucrin; neural tissue-specific cysteine-rich protein; PSEC0258; UNQ3119; UNQ3119/PRO10268
Gene Symbols DRAXIN
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Draxin Gly26-Ile343.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 691317, 70433
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.