Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRB1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP187966
Description
DRB1 Polyclonal specifically detects DRB1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
DRB1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
RBM45 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VDSLDEPPNSRIFLVISKYTPESVLRERFSPFGDIQDIWVVRDKHTKESKGIAFVKFARSSQACRAMEEMHGQCLGPNDTKPIKVFIAQSRSSGSHRDVEDEELTRIFVMIPKSYTEE | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.1mg/mL | |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
developmentally regulated RNA-binding protein 1, Developmentally-regulated RNA-binding protein 1, DRB1FLJ44612, DRBP1, MGC42237, putative RNA binding protein RB-1, RB-1, RNA binding motif protein 45, RNA-binding motif protein 45, RNA-binding protein 45 | |
Rabbit | |
Affinity Purified | |
RUO | |
129831 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction