Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DRB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541951
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541951 Supplier Invitrogen™ Supplier No. PA541951
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MDEAGSSASG GGFRPGVDSL DEPPNSRIFL VISKYTPESV LRERFSPFGD Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

DRB1 is RNA-binding protein with binding specificity for poly(C) and may play an important role in neural development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRB1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RBM45
Gene Accession No. Q8IUH3
Gene Alias developmentally regulated RNA binding protein 1; developmentally regulated RNA-binding protein 1; developmentally-regulated RNA-binding protein 1; dopamine receptor binding 1; Drb1; Drbp1; G430095G15Rik; putative RNA binding protein RB-1; RB-1; Rbm45; RNA binding motif protein 45; RNA-binding motif protein 45; RNA-binding protein 45
Gene Symbols RBM45
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DRB1 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129831
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.