Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DRB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589477
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589477 Supplier Invitrogen™ Supplier No. PA589477
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDEAGSSASG GGFRPGVDSL DEPPNSRIFL VISKYTPESV LRERFSPFGD IQDIWVVRDK HTKESKGIAF VKFARSSQAC RAMEEMHGQC LGPNDTKPIK VFIAQSRSSG SHRDVEDEEL TRIFVMIPKS YTEEDLREKF KVYGDIEYCS IIKNKVTGES KGLGYVRYLK PSQAAQAIEN CDRSFRAILA EPKNKASESS EQDYYSNMRQ EALGHEPRVN MFPFEQQSEF SSFDKNDSRG QEAISKRLSV VSRVPFTEEQ LFSIFDIVPG LEYCEVQRDP YSNYGHGVVQ YFNVASAIYA; Positive Samples: Mouse brain, Mouse heart, Mouse kidney; Cellular Location: Cytoplasm, Nucleus.

DRB1 is RNA-binding protein with binding specificity for poly(C) and may play an important role in neural development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRB1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RBM45
Gene Accession No. Q8BHN5
Gene Alias developmentally regulated RNA binding protein 1; developmentally regulated RNA-binding protein 1; developmentally-regulated RNA-binding protein 1; dopamine receptor binding 1; Drb1; Drbp1; G430095G15Rik; putative RNA binding protein RB-1; RB-1; Rbm45; RNA binding motif protein 45; RNA-binding motif protein 45; RNA-binding protein 45
Gene Symbols RBM45
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RBM45 (NP_6944532).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241490
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.