Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DRB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113111
Description
DRB1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
DRB1 is RNA-binding protein with binding specificity for poly(C) and may play an important role in neural development.
Specifications
| DRB1 | |
| Polyclonal | |
| Unconjugated | |
| RBM45 | |
| developmentally regulated RNA binding protein 1; developmentally regulated RNA-binding protein 1; developmentally-regulated RNA-binding protein 1; dopamine receptor binding 1; Drb1; Drbp1; G430095G15Rik; putative RNA binding protein RB-1; RB-1; Rbm45; RNA binding motif protein 45; RNA-binding motif protein 45; RNA-binding protein 45 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 129831, 266631 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 3.02 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8CFD1, Q8IUH3 | |
| RBM45 | |
| Recombinant Human RNA-binding protein 45 protein (170-244AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction