Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRD1 Monoclonal Antibody (SG2-D1a), Invitrogen™

Catalog No. MA146024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA146024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA146024 Supplier Thermo Scientific Supplier No. MA146024

Mouse Monoclonal Antibody

This antibody does not react with human, canine, porcine, sheep, rabbit, or bovine.

The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRD1
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone SG2-D1a
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DRD1
Gene Accession No. P18901, Q61616
Gene Alias C030036C15Rik; D(1A) dopamine receptor; D1 receptor; D1a; D1A dopamine receptor; D1a receptor; DADR; Dopamine 1 receptor; Dopamine D1 receptor; dopamine receptor D1; dopamine receptor D1A; Dopamine-1A receptor; Drd1; Drd-1; Drd1a; G protein-coupled receptor DRD1; Gpcr15
Gene Symbols DRD1
Host Species Mouse
Immunogen Recombinant rat D1a dopamine receptor (123 residues with 23 residues near the C-terminus).[Swiss-Prot# P18901]
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13488, 24316
Target Species Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.