Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRD1 Rabbit anti-Human, Mouse, Unconjugated, Clone: AAEA-4, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19988
Description
DRD1 Monoclonal antibody specifically detects DRD1 in Human, Mouse samples. It is validated for WB-Ce.Specifications
DRD1 | |
Monoclonal | |
Unconjugated | |
DRD1 | |
Rabbit | |
100 μg | |
1812 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
AAEA-4 | |
dopamine receptor D1 | |
DADR, DRD1A | |
A synthetic peptide corresponding to human DRD1 | |
Primary | |
Human, Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction