Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DRD1 Recombinant Rabbit Monoclonal Antibody (JM10-93)
GREENER_CHOICE

Catalog No. PIMA542467
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542467 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542467 Supplier Invitrogen™ Supplier No. MA542467
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: Mouse kidney tissue lysates, 293T, N2A, SH-SY5Y, mouse brain tissue, rat brain tissue. Subcellular Location: Cell membrane. Endoplasmic reticulum membrane.

The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JM10-93
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene DRD1
Gene Accession No. P18901, P21728, Q61616
Gene Alias C030036C15Rik; D(1A) dopamine receptor; D1 receptor; D1a; D1A dopamine receptor; D1a receptor; DADR; Dopamine 1 receptor; Dopamine D1 receptor; dopamine receptor D1; dopamine receptor D1A; Dopamine-1A receptor; Drd1; Drd-1; Drd1a; G protein-coupled receptor DRD1; Gpcr15
Gene Symbols DRD1
Host Species Rabbit
Immunogen Synthetic peptide within C-terminal human DRD1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13488, 1812, 24316
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.