Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DRD2 Recombinant Rabbit Monoclonal Antibody (HL1584)
GREENER_CHOICE

Catalog No. PIMA547103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547103 Supplier Invitrogen™ Supplier No. MA547103
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The DRD2 Dopamine Receptor is involved in the neuromodulation of appetitive behaviors. DRD2 may be linked to alcoholism and other substance-use disorders. Mutations in DRD2 in mice lead to the suppression of reward behavior with morphine and to the reduction of spontaneous movements, a phenotype resembling Parkinsons disease. Three alternatively spliced isoforms have been identified. DRD2 has been reported to be expressed in various regions of the brain, as well as in aorta, artery, eye, and spinal cord. ESTs have been isolated from normal pituitary and in brain, colon, lung, pancreas, and skeletal muscle cancer libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRD2
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone HL1584
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DRD2
Gene Accession No. P14416, P61168, P61169
Gene Alias D(2) dopamine receptor; D2 receptor; D2DR; D2R; Dopamine 2 receptor; Dopamine D2 receptor; dopamine receptor 2; dopamine receptor D2; dopamine receptor D2 isoform; Drd2; Drd-2; seven transmembrane helix receptor
Gene Symbols DRD2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human Dopamine Receptor D2. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13489, 1813, 24318
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.