Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 220221AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-114-2714


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

The DRD2 Dopamine Receptor is involved in the neuromodulation of appetitive behaviors. DRD2 may be linked to alcoholism and other substance-use disorders. Mutations in DRD2 in mice lead to the suppression of reward behavior with morphine and to the reduction of spontaneous movements, a phenotype resembling Parkinsons disease. Three alternatively spliced isoforms have been identified. DRD2 has been reported to be expressed in various regions of the brain, as well as in aorta, artery, eye, and spinal cord. ESTs have been isolated from normal pituitary and in brain, colon, lung, pancreas, and skeletal muscle cancer libraries.
Specifications

Specifications

DRD2
Polyclonal
Unconjugated
DRD2
D(2) dopamine receptor, D2DR, D2R, Dopamine D2 receptor, dopamine receptor D2, DRD2
Rabbit
Antigen Affinity Chromatography
RUO
13489, 1813
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P14416, P61168
DRD2
DRD2 Fusion Protein Ag16821
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.