Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRD5/Dopamine receptor 5 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1747R

Description
The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.Specifications
DRD5/Dopamine receptor 5 | |
Polyclonal | |
Unconjugated | |
DRD5 | |
D(1B) dopamine receptor; D(5) dopamine receptor; D1B; D1b receptor; D1beta dopamine receptor; D5R; DBDR; Dopamine 5 receptor; Dopamine D5 receptor; dopamine receptor 5; dopamine receptor D1B; dopamine receptor D5; DRD 1B; DRD 5; DRD1B; DRD1L2; DRD5; Drd-5; Gpcr1; Hd5dr; MGC10601 | |
Rabbit | |
Protein A | |
RUO | |
1816 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P21918 | |
DRD5 | |
KLH conjugated synthetic peptide derived from rat DRD5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction