Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DreamTaq DNA Polymerase (5 U/μL)

Thermo Scientific DreamTaq DNA Polymerase is an enhanced Taq DNA polymerase optimized for all standard PCR applications. It ensures higher sensitivity, longer PCR products, and higher yields compared to conventional Taq DNA polymerase.

Supplier:  Thermo Scientific™ EP0712

 View more versions of this product

Catalog No. FEREP0712


Only null left
Add to Cart

Description

Description

Thermo Scientific DreamTaq DNA Polymerase is an enhanced Taq DNA polymerase optimized for all standard PCR applications. It ensures higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase. DreamTaq DNA Polymerase uses the same reaction setup and cycling conditions as conventional Taq DNA polymerase. Extensive optimization of reaction conditions is not required. It is supplied with optimized DreamTaq Buffer, which includes 20 mM MgCl2

DreamTaq Green DNA Polymerase is a combination of DreamTaq DNA Polymerase and 10X DreamTaq Green Buffer. The 10X DreamTaq Green Buffer includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with PCR performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Features

  • Robust amplification with minimal optimization
  • High yields of PCR products
  • Higher sensitivity compared to conventional Taq DNA polymerase
  • Amplification of long targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
  • Incorporates modified nucleotides, but does not incorporate dUTP
  • Direct loading of PCR product on gel with green format

Applications

  • Routine PCR amplification of DNA fragments up to 6 kb
  • High throughput PCR
  • Genotyping

Notes

  • For applications requiring PCR product analysis by absorbance or fluorescence excitation, colorless 10X DreamTaq Buffer (Cat. No. B65) is recommended for purifying PCR products before analysis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 U/μL
Low
Standard
Standard PCR
No
Separate Components
• DreamTaq DNA Polymerase (5 U/μL), 100 μL
• 10X DreamTaq Green Buffer with 20 mM MgCl2, 2 x 1.25 mL

Store at -20°C.
DreamTaq DNA Polymerase
500 Units
1X
3'-A
Green
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.