Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) PROMO
Save time in PCR applications with Master Mix and get higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase.
Supplier: Thermo Scientific™ K1082
Description
Highlights
- Direct loading of PCR product on gel
- High yields and high sensitivity of PCR
- Amplification of long targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
- Robust amplification with minimal optimization
- Incorporates modified nucleotides, but does not incorporate dUTP
Applications
- Routine PCR amplification of DNA fragments up to 6 kb
- Genotyping
- High throughput PCR
Specifications
Contains: 20 x 1.25 mL DreamTaq Green PCR Master Mix (2X), which includes DreamTaq DNA Polymerase, 2X DreamTaq Green buffer, dNTPs, and 4 mM MgCl2, 20 x 1.25 mL Nuclease-free water , Store at -20°C. | |
DreamTaq DNA Polymerase | |
DreamTaq™ | |
1X | |
3'-A |
Low | |
Standard | |
1000 Reactions | |
No | |
SuperMix or Master Mix |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction