Learn More
Drebrin 1 Antibody (DBN-N-03), Alexa Fluor™ 594, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP222344AF594
Catalog No. NP222344A94
Only null left
Add to Cart
Description
Description
Drebrin 1 Monoclonal specifically detects Drebrin 1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
Drebrin 1
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
DBN1
Bacterially expressed N-terminal fragment of recombinant human Drebrin Antibody (DBN-N-03)
Primary
The mouse monoclonal Drebrin Antibody (DBN-N-03) recognizes drebrin, an approximately 100-125 kDa regulator of actin cytoskeleton.
Store at 4°C in the dark.
Western Blot, Flow Cytometry
DBN-N-03
Western Blot, Flow Cytometry
D0S117E, Developmentally-regulated brain protein, DKFZp434D064, drebrin, drebrin 1, drebrin E, drebrin E2, drebrin-1
Mouse
0.1 mL
1627
Human
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.