Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Drebrin 1 Antibody (DBN-N-03), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222344AF647
Description
Drebrin 1 Monoclonal specifically detects Drebrin 1 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
Drebrin 1 | |
Monoclonal | |
Alexa Fluor 647 | |
D0S117E, Developmentally-regulated brain protein, DKFZp434D064, drebrin, drebrin 1, drebrin E, drebrin E2, drebrin-1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The mouse monoclonal Drebrin Antibody (DBN-N-03) recognizes drebrin, an approximately 100-125 kDa regulator of actin cytoskeleton. | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry | |
DBN-N-03 | |
Western Blot, Flow Cytometry | |
DBN1 | |
Bacterially expressed N-terminal fragment of recombinant human Drebrin Antibody (DBN-N-03) | |
0.1 mL | |
Neuroscience | |
1627 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction