Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Drebrin 1 Antibody (DBN-N-03), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222344MFV610
Description
Drebrin 1 Monoclonal antibody specifically detects Drebrin 1 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
Drebrin 1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
DBN-N-03 | |
Flow Cytometry, Flow (Intracellular) | |
D0S117E, Developmentally-regulated brain protein, DKFZp434D064, drebrin, drebrin 1, drebrin E, drebrin E2, drebrin-1 | |
Bacterially expressed N-terminal fragment of recombinant human Drebrin Antibody (DBN-N-03) | |
0.1 mL | |
Neuroscience | |
1627 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction