Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Drebrin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100142
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100142 Supplier Invitrogen™ Supplier No. PA5100142
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Drebrin.

The protein encoded by this gene is a cytoplasmic actin-binding protein thought to play a role in the process of neuronal growth. It is a member of the drebrin family of proteins that are developmentally regulated in the brain. A decrease in the amount of this protein in the brain has been implicated as a possible contributing factor in the pathogenesis of memory disturbance in Alzheimer's disease. At least two alternative splice variants encoding different protein isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Drebrin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DBN1
Gene Accession No. Q07266, Q16643, Q9QXS6
Gene Alias D0S117E; Dbn1; developmentally regulated brain protein; developmentally-regulated brain protein; DKFZp434D064; Drba; Drebrin; drebrin 1; drebrin A; drebrin E; drebrin E2; drebrin-1; Unknown (protein for MGC:138067)
Gene Symbols DBN1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DBN1(Accession Q16643), corresponding to amino acid residues S304-R354.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1627, 56320, 81653
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.