Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DREF Antibody (OTI5A2), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271419V
Description
DREF Monoclonal specifically detects DREF in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
DREF | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZBED1 | |
Full length recombinant protein of human DREF (NP_004720) produced in HEK293T cells. | |
0.1 mL | |
9189.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI5A2 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Ac-like transposable element, ALTEDNA replication-related element binding factor, hDREF, KIAA0785dREF homolog, Putative Ac-like transposable element, putative Ac-like transposon, TRAMPDREFBED-type zinc finger domain-containing protein 1, zinc finger BED domain-containing protein 1, zinc finger, BED-type containing 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction