Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DREF Antibody (OTI5A2), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271419PCP
Description
DREF Monoclonal specifically detects DREF in Human samples. It is validated for Flow Cytometry.Specifications
| DREF | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| ZBED1 | |
| Full length recombinant protein of human DREF (NP_004720) produced in HEK293T cells. | |
| 0.1 mL | |
| 9189.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI5A2 | |
| Flow Cytometry | |
| Ac-like transposable element, ALTEDNA replication-related element binding factor, hDREF, KIAA0785dREF homolog, Putative Ac-like transposable element, putative Ac-like transposon, TRAMPDREFBED-type zinc finger domain-containing protein 1, zinc finger BED domain-containing protein 1, zinc finger, BED-type containing 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction